2-Fluor-5-nitrobenzonitril

Lieferant: COMBI-BLOCKS

AN-1040-25G AN-1040-100G
COBBAN-1040-25GEA 58.6 EUR
COBBAN-1040-25G COBBAN-1040-100G
2-Fluor-5-nitrobenzonitril
2-Fluor-5-nitrobenzonitril
Formel: C₇H₃FN₂O₂
Molecular Weight: 166,11 g/mol
Schmelzpunkt: 79 °C
MDL: MFCD00042299
CAS-Nummer: 17417-09-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR