2-Fluor-6-methoxybenzonitril

Lieferant: COMBI-BLOCKS

OS-7506-25G OS-7506-100G
COBBOS-7506-25GEA 42.3 EUR
COBBOS-7506-25G COBBOS-7506-100G
2-Fluor-6-methoxybenzonitril
2-Fluor-6-methoxybenzonitril
Formel: C₈H₆FNO
Molecular Weight: 151.14 g/mol
Siedepunkt: 245
Schmelzpunkt: 93…95 °C
MDL: MFCD00042291
CAS-Nummer: 94088-46-7
EINECS: 302-047-5
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR