3,4-Difluor-2-hydroxybenzonitril

Lieferant: CHEMPUR

FL13817.POR 013817-1G
CHMPFL13817.POREA 0 EUR
CHMPFL13817.POR FLUO013817-1G
3,4-Difluor-2-hydroxybenzonitril
3,4-Difluor-2-hydroxybenzonitril
Formel: C₇H₃F₂NO
Molecular Weight: 155.1 g/mol
CAS-Nummer: 186590-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR