3,4-Difluor-2-methoxybenzonitril

Lieferant: CHEMPUR

Synonyme: 3,4-Difluor-2-methoxybenzoesäurenitril

FL13821.POR 013821-1G 013821-5G
CHMPFL13821.POREA 0 EUR
CHMPFL13821.POR FLUO013821-1G FLUO013821-5G
3,4-Difluor-2-methoxybenzonitril
3,4-Difluor-2-methoxybenzonitril
Formel: C₈H₅F₂NO
Molecular Weight: 169,13 g/mol
MDL: MFCD03428594
CAS-Nummer: 886496-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR