4-Hydroxy-3,5-diiodbenzonitril
Lieferant: CHEMPUR
Synonyme:
Ioxynil
SPB00182.250
SPB00182.1
CHMPSPB00182.250EA
0
EUR
CHMPSPB00182.250
MAYBSPB00182.1
4-Hydroxy-3,5-diiodbenzonitril
4-Hydroxy-3,5-diiodbenzonitril
Formel:
C₇H₃I₂NO Molecular Weight: 370,92 g/mol Siedepunkt: 378 °C (1013 hPa) Schmelzpunkt: 212 °C |
MDL:
MFCD00016422 CAS-Nummer: 1689-83-4 EINECS: 216-881-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...