3-Fluor-o-toluonitril
Lieferant: COMBI-BLOCKS
OS-7481-5G
OS-7481-25G
OS-7481-1G
COBBOS-7481-5GEA
101
EUR
COBBOS-7481-5G
COBBOS-7481-25G
COBBOS-7481-1G
3-Fluor-o-toluonitril
3-Fluor-o-toluonitril
Formel:
C₈H₆FN Molecular Weight: 135.14 g/mol |
MDL:
MFCD03788541 CAS-Nummer: 185147-06-2 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...