3-Fluor-4-hydroxybenzonitril

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluor-4-hydroxybenzoesäurenitril

CA-4263-5G CA-4263-25G CA-4263-100G
COBBCA-4263-5GEA 0 EUR
COBBCA-4263-5G COBBCA-4263-25G COBBCA-4263-100G
3-Fluor-4-hydroxybenzonitril
3-Fluor-4-hydroxybenzonitril
Formel: C₇H₄FNO
Molecular Weight: 137.11 g/mol
MDL: MFCD00215834
CAS-Nummer: 405-04-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR