3-Fluor-5-methoxybenzonitril
Lieferant: COMBI-BLOCKS
Synonyme:
3-Fluor-5-methoxybenzoesäurenitril
CA-4151-5G
CA-4151-1G
COBBCA-4151-5GEA
119
EUR
COBBCA-4151-5G
COBBCA-4151-1G
3-Fluor-5-methoxybenzonitril
3-Fluor-5-methoxybenzonitril
Formel:
C₈H₆FNO Molecular Weight: 151.14 g/mol |
MDL:
MFCD06797883 CAS-Nummer: 439280-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...