4-(Trifluormethoxy)benzonitril
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Trifluormethoxy)benzoesäurenitril
OS-7672-5G
OS-7672-100G
OS-7672-25G
COBBOS-7672-5GEA
25.5
EUR
COBBOS-7672-5G
COBBOS-7672-100G
COBBOS-7672-25G
4-(Trifluormethoxy)benzonitril
4-(Trifluormethoxy)benzonitril
Formel:
C₈H₄F₃NO Molecular Weight: 187.12 g/mol Siedepunkt: 192 Dichte: 1.284 g/cm³ (20 °C) |
MDL:
MFCD00039474 CAS-Nummer: 332-25-2 EINECS: 206-363-3 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...