4-Fluor-o-toluonitril

Lieferant: COMBI-BLOCKS

HC-6859-5G HC-6859-25G HC-6859-1G
COBBHC-6859-5GEA 0 EUR
COBBHC-6859-5G COBBHC-6859-25G COBBHC-6859-1G
4-Fluor-o-toluonitril
4-Fluor-o-toluonitril
Formel: C₈H₆FN
Molecular Weight: 135.14 g/mol
MDL: MFCD03095106
CAS-Nummer: 147754-12-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR