4-Fluor-3-methoxybenzonitril

Lieferant: FLUOROCHEM

Synonyme: 4-Fluor-3-methoxybenzoesäurenitril

CFF1127. 013922-5G 013922-1G
CHMPCFF1127.EA 0 EUR
CHMPCFF1127. FLUO013922-5G FLUO013922-1G
4-Fluor-3-methoxybenzonitril
4-Fluor-3-methoxybenzonitril
Formel: C₈H₆FNO
Molecular Weight: 151.14 g/mol
MDL: MFCD01569371
CAS-Nummer: 243128-37-2
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR