4-Fluor-m-toluonitril

Lieferant: COMBI-BLOCKS

HC-6964-5G HC-6964-25G HC-6964-100G
COBBHC-6964-5GEA 26.2 EUR
COBBHC-6964-5G COBBHC-6964-25G COBBHC-6964-100G
4-Fluor-m-toluonitril
4-Fluor-m-toluonitril
Formel: C₈H₆FN
Molecular Weight: 135,14 g/mol
MDL: MFCD03788510
CAS-Nummer: 185147-08-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR