4-Nitrobenzonitril
Lieferant: COMBI-BLOCKS
|
Gefahr
|
Synonyme:
4-Nitrobenzoesäurenitril
AN-1261-25G
AN-1261-1G
AN-1261-100G
COBBAN-1261-25GEA
0
EUR
COBBAN-1261-25G
COBBAN-1261-1G
COBBAN-1261-100G
4-Nitrobenzonitril
4-Nitrobenzonitril
Formel:
C₇H₄N₂O₂ Molecular Weight: 148,12 g/mol Schmelzpunkt: 145…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007279 CAS-Nummer: 619-72-7 EINECS: 210-610-0 UN: 3439 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...