3'-Fluor-4'-methoxyacetophenon

Lieferant: BIONET RESEARCH

Synonyme: 1-(3-Fluoro-4-methoxyphenyl)-1-ethanone

7Y-0947.1 7Y-0947.10 7Y-0947.5 7Y-0947.500
BIOT7Y-0947.1EA 0 EUR
BIOT7Y-0947.1 BIOT7Y-0947.10 BIOT7Y-0947.5 BIOT7Y-0947.500
3'-Fluor-4'-methoxyacetophenon
3'-Fluor-4'-methoxyacetophenon
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
Siedepunkt: 147…148 °C (20 mmHg)
Schmelzpunkt: 92 °C
MDL: MFCD00026219
CAS-Nummer: 455-91-4
EINECS: 207-253-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR