4'-Isobutylacetophenon ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-(4-Isobutylphenyl)ethanon, 4-Isobutyl-1-acetylbenzol, p-Isobutylacetophenon
A10624.18
A10624.30
A10624.18EA
60.5
EUR
A10624.18
A10624.30
4'-Isobutylacetophenon ≥97%
4'-Isobutylacetophenon
Purity: 97%
CAS No.: 38861-78-8
Molecular Formula: C12H16O
Formula Weight: 176.27
Flash Point: 54°(129°F)
Refractive Index: 1.5180
Beilstein Reference: 1935275
UN No.: 1224
MDL No.: MFCD00027393
Formel:
C₁₂H₁₆O Molecular Weight: 176.26 Dichte: 0.952 Flammpunkt: 54 °C (129 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00027393 CAS-Nummer: 38861-78-8 EINECS: 254-159-8 UN: 1224 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...