2-Methoxy-4-vinylphenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-5-vinylanisol

M-Clarity™ Qualitätsstufe = MQ300

W267511-100G-K W267511-1KG-K W267511-25G-K
SIALW267511-100G-KEA 632 EUR
SIALW267511-100G-K SIALW267511-1KG-K SIALW267511-25G-K
2-Methoxy-4-vinylphenol, Sigma-Aldrich®
2-Methoxy-4-vinylphenol
Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
MDL: MFCD00015437
CAS-Nummer: 7786-61-0
EINECS: 232-101-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR