3-Ethyl-2-iodtoluol

Lieferant: abcr

Synonyme: 1-Ethyl-2-iod-3-methylbenzol, 2-Ethyl-6-methyliodbenzol

10839 10839.0100
ABCR10839EA 0 EUR
ABCR10839 ABCR10839.0100
3-Ethyl-2-iodtoluol
3-Ethyl-2-iodtoluol
Formel: C₉H₁₁I
Molecular Weight: 246.09 g/mol
MDL: MFCD00052855
CAS-Nummer: 175277-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR