o,α,α,α-Tetrafluoranisol

Lieferant: COMBI-BLOCKS

Synonyme: α,α,α,2-Tetrafluoranisol, 1-Fluor-2-(trifluormethoxy)benzol, 2-(Trifluormethoxy)fluorbenzol

OS-7599-25G OS-7599-5G
COBBOS-7599-25GEA 0 EUR
COBBOS-7599-25G COBBOS-7599-5G
o,α,α,α-Tetrafluoranisol
o,α,α,α-Tetrafluoranisol
Formel: C₇H₄F₄O
Molecular Weight: 180.1 g/mol
MDL: MFCD00236322
CAS-Nummer: 2106-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR