2-Fluor-3-(trifluormethyl)phenol
Lieferant: FLUOROCHEM
Synonyme:
2-Fluoro-3-hydroxybenzotrifluoride
006429-1G
006429-5G
FLUO006429-1GEA
148
EUR
FLUO006429-1G
FLUO006429-5G
2-Fluor-3-(trifluormethyl)phenol
2-Fluor-3-(trifluormethyl)phenol
Formel:
C₇H₄F₄O Molecular Weight: 180,1 g/mol |
MDL:
MFCD00083554 CAS-Nummer: 207291-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...