2-Fluor-4-nitroanisol

Lieferant: COMBI-BLOCKS
Achtung

AN-1093-5G AN-1093-25G AN-1093-100G
COBBAN-1093-5GEA 0 EUR
COBBAN-1093-5G COBBAN-1093-25G COBBAN-1093-100G
2-Fluor-4-nitroanisol
2-Fluor-4-nitroanisol
Formel: C₇H₆FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00061095
CAS-Nummer: 455-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR