3-Fluor-2-methoxyphenol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluor-6-hydroxyanisol, 1-Fluor-3-hydroxy-2-methoxybenzol
OT-0938-5G
OT-0938-1G
COBBOT-0938-5GEA
1490
EUR
COBBOT-0938-5G
COBBOT-0938-1G
3-Fluor-2-methoxyphenol
3-Fluor-2-methoxyphenol
Formel:
C₇H₇FO₂ Molecular Weight: 142.13 g/mol |
MDL:
MFCD12756463 CAS-Nummer: 96994-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...