5-Fluor-2-methoxyphenol
Lieferant: CHEMPUR
Synonyme:
4-Fluor-2-hydroxyanisol, 1-Fluor-3-hydroxy-4-methoxybenzol
FL13351.POR
013351-25G
013351-5G
CHMPFL13351.POREA
0
EUR
CHMPFL13351.POR
FLUO013351-25G
FLUO013351-5G
5-Fluor-2-methoxyphenol
5-Fluor-2-methoxyphenol
Formel:
C₇H₇FO₂ |
MDL:
MFCD00143459 CAS-Nummer: 72955-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...