3-Fluor-5-(trifluormethyl)phenol

Lieferant: FLUOROCHEM

Synonyme: 3-Fluor-5-hydroxybenzotrifluorid

006432-1G 006432-5G 006432-25G
FLUO006432-1GEA 115 EUR
FLUO006432-1G FLUO006432-5G FLUO006432-25G
3-Fluor-5-(trifluormethyl)phenol
3-Fluor-5-(trifluormethyl)phenol
Formel: C₇H₄F₄O
Molecular Weight: 180.1 g/mol
MDL: MFCD00061288
CAS-Nummer: 172333-87-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR