5-Fluor-2-nitroanisol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-2-methoxy-1-nitrobenzene

AN-1018-5G AN-1018-25G AN-1018-100G
COBBAN-1018-5GEA 0 EUR
COBBAN-1018-5G COBBAN-1018-25G COBBAN-1018-100G
5-Fluor-2-nitroanisol
5-Fluor-2-nitroanisol
Formel: C₇H₆FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00077541
CAS-Nummer: 448-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR