3-Fluor-4-nitroanisol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-4-methoxy-1-nitrobenzene
AN-1026-5G
AN-1026-25G
AN-1026-1G
COBBAN-1026-5GEA
0
EUR
COBBAN-1026-5G
COBBAN-1026-25G
COBBAN-1026-1G
3-Fluor-4-nitroanisol
3-Fluor-4-nitroanisol
Formel:
C₇H₆FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115632 CAS-Nummer: 446-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...