4-Fluor-2-nitroanisol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 4-Fluoro-1-methoxy-2-nitrobenzene

AN-1148-100G AN-1148-25G AN-1148-5G
COBBAN-1148-100GEA 352 EUR
COBBAN-1148-100G COBBAN-1148-25G COBBAN-1148-5G
4-Fluor-2-nitroanisol
4-Fluor-2-nitroanisol
Formel: C₇H₆FNO₃
Molecular Weight: 171,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013375
CAS-Nummer: 445-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR