4-Fluor-3-nitrotoluol

Lieferant: COMBI-BLOCKS

AN-1103-5G AN-1103-25G AN-1103-100G
COBBAN-1103-5GEA 0 EUR
COBBAN-1103-5G COBBAN-1103-25G COBBAN-1103-100G
4-Fluor-3-nitrotoluol
4-Fluor-3-nitrotoluol
Formel: C₇H₆FNO₂
MDL: MFCD00007060
CAS-Nummer: 446-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR