3-(Trifluormethoxy)phenol
Lieferant: COMBI-BLOCKS
Synonyme:
α,α,α-Trifluor-3-hydroxyanisol
OS-7670-5G
OS-7670-25G
OS-7670-1G
COBBOS-7670-5GEA
38.5
EUR
COBBOS-7670-5G
COBBOS-7670-25G
COBBOS-7670-1G
3-(Trifluormethoxy)phenol
3-(Trifluormethoxy)phenol
Formel:
C₇H₅F₃O₂ Molecular Weight: 178,11 g/mol Siedepunkt: 69,5…79.5 °C (24 mmHg) Dichte: 1,37 g/cm³ (20 °C) |
MDL:
MFCD00040987 CAS-Nummer: 827-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...