3-Nitro-5-(trifluormethyl)phenol

Lieferant: CHEMPUR

Synonyme: 3-Hydroxy-5-nitrobenzotrifluorid

FL13855.POR 013855-25G 013855-5G 013855-1G
CHMPFL13855.POREA 0 EUR
CHMPFL13855.POR FLUO013855-25G FLUO013855-5G FLUO013855-1G
3-Nitro-5-(trifluormethyl)phenol
3-Nitro-5-(trifluormethyl)phenol
Formel: C₇H₄F₃NO₃
Molecular Weight: 207,11 g/mol
MDL: MFCD04973779
CAS-Nummer: 349-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR