2-Iod-4-nitroanisol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Iodo-1-methoxy-4-nitrobenzene
HC-3364-5G
HC-3364-25G
COBBHC-3364-5GEA
38.5
EUR
COBBHC-3364-5G
COBBHC-3364-25G
2-Iod-4-nitroanisol
2-Iod-4-nitroanisol
Formel:
C₇H₆INO₃ Molecular Weight: 279,03 g/mol |
MDL:
MFCD00024328 CAS-Nummer: 5399-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...