2-Iod-5-nitrotoluol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 1-Iodo-2-methyl-4-nitrobenzene

HC-3050-25G HC-3050-1G HC-3050-5G
COBBHC-3050-25GEA 102 EUR
COBBHC-3050-25G COBBHC-3050-1G COBBHC-3050-5G
2-Iod-5-nitrotoluol
2-Iod-5-nitrotoluol
Formel: C₇H₆INO₂
Molecular Weight: 263,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051517
CAS-Nummer: 5326-38-5
EINECS: 226-204-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR