5-Methyl-2-nitroanisol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 1-Methoxy-3-methyl-6-nitrobenzol, 3-Methoxy-4-nitrotoluol, Methyl-6-nitro-m-tolylether

AN-1146-5G AN-1146-100G AN-1146-25G
COBBAN-1146-5GEA 0 EUR
COBBAN-1146-5G COBBAN-1146-100G COBBAN-1146-25G
5-Methyl-2-nitroanisol
5-Methyl-2-nitroanisol
Formel: C₈H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00007367
CAS-Nummer: 38512-82-2
EINECS: 253-975-1
UN: 2446
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR