1-Methylindol
Lieferant: COMBI-BLOCKS
Synonyme:
N-Methylindole, 1-Methyl-1H-indole
IN-0166-100G
IN-0166-5G
IN-0166-25G
COBBIN-0166-100GEA
154
EUR
COBBIN-0166-100G
COBBIN-0166-5G
COBBIN-0166-25G
1-Methylindol
1-Methylindol
Formel:
C₉H₉N Molecular Weight: 131.18 g/mol Siedepunkt: 239 Schmelzpunkt: 94…95 °C Dichte: 1.048 g/cm³ (20 °C) |
MDL:
MFCD00005800 CAS-Nummer: 603-76-9 EINECS: 210-057-5 UN: 3334 ADR: 9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...