1,2,3-Trifluor-5-nitrobenzol, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
3,4,5-Trifluornitrobenzol
M-Clarity™ Qualitätsstufe = MQ100
656917-25G
656917-5G
SIAL656917-25GEA
611
EUR
SIAL656917-25G
SIAL656917-5G
1,2,3-Trifluor-5-nitrobenzol, Sigma-Aldrich®
1,2,3-Trifluor-5-nitrobenzol
Formel:
C₆H₂F₃NO₂ Molecular Weight: 177,08 g/mol |
MDL:
MFCD00456803 CAS-Nummer: 66684-58-0 EINECS: 266-447-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...