1-Undecen ≥97%

Lieferant: Thermo Scientific
Achtung

H53387.06 H53387.14
H53387.06EA 80.8 EUR
H53387.06 H53387.14
1-Undecen ≥97%
1-Undecen

Purity: 97% CAS No.: 821-95-4 Molecular Formula: C11H22 Formula Weight: 154.30 Flash Point: 63°(145°F) Refractive Index: 1.426 Melting Point: -49° Boiling Point: 192-193° Beilstein Reference: 1740044 MDL No.: MFCD00008956

Formel: C₁₁H₂₂
Molecular Weight: 154,3 g/mol
Siedepunkt: 192…193 °C
Schmelzpunkt: –49 °C
Dichte: 0,751
Flammpunkt: 63 °C (145 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008956
CAS-Nummer: 821-95-4
EINECS: 212-483-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR