4'-Hydroxy-6-methylflavon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Methyl-2-(4-hydroxyphenyl)-4-benzopyron, 6-Methyl-2-(4-hydroxyphenyl)chromen-4-on, 6-Methyl-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-on
H30959.03
H30959.06
H30959.14
H30959.03EA
24.2
EUR
H30959.03
H30959.06
H30959.14
4'-Hydroxy-6-methylflavon ≥97%
4'-Hydroxy-6-methylflavon
Purity: 97%
CAS No.: 288401-04-7
Molecular Formula: C16H12O3
Formula Weight: 252.28
Melting Point: 238-243°
MDL No.: MFCD03424432
Formel:
C₁₆H₁₂O₃ Molecular Weight: 252,27 g/mol Schmelzpunkt: 238…243 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424432 CAS-Nummer: 288401-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...