2-(Chlormethyl)benzimidazol

Lieferant: BIONET RESEARCH

Synonyme: 2-(Chlormethyl)-1H-benzimidazol

7J-352S.500 7J-352S.1 7J-352S.5 7J-352S.10
BIOT7J-352S.500EA 0 EUR
BIOT7J-352S.500 BIOT7J-352S.1 BIOT7J-352S.5 BIOT7J-352S.10
2-(Chlormethyl)benzimidazol
2-(Chloromethyl)benzimidazole
Formel: C₈H₇ClN₂
MDL: MFCD00005602
CAS-Nummer: 4857-04-9
EINECS: 225-452-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR