2-(Chlormethyl)benzimidazol
Lieferant: FLUOROCHEM
Synonyme:
2-(Chlormethyl)-1H-benzimidazol
010651-5G
010651-25G
FLUO010651-5GEA
42.9
EUR
FLUO010651-5G
FLUO010651-25G
2-(Chlormethyl)benzimidazol
2-(Chloromethyl)benzimidazole
Formel:
C₈H₇ClN₂ Molecular Weight: 166,61 g/mol |
MDL:
MFCD00005602 CAS-Nummer: 4857-04-9 EINECS: 225-452-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...