2-(Chlormethyl)benzimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Chlormethyl)-1H-benzimidazol

C52055-25G C52055-5G
SIALC52055-25GEA 100 EUR
SIALC52055-25G SIALC52055-5G
2-(Chlormethyl)benzimidazol, Sigma-Aldrich®
2-(Chloromethyl)benzimidazole
Formel: C₈H₇ClN₂
Molecular Weight: 166,61 g/mol
MDL: MFCD00005602
CAS-Nummer: 4857-04-9
EINECS: 225-452-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR