2-(Diphenylphosphino)benzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

454885-1G 454885-5G
SIAL454885-1GEA 184 EUR
SIAL454885-1G SIAL454885-5G
2-(Diphenylphosphino)benzoesäure, Sigma-Aldrich®
2-(Diphenylphosphino)benzoic acid
Formel: C₁₉H₁₅O₂P
Molecular Weight: 306,3 g/mol
MDL: MFCD00674024
CAS-Nummer: 17261-28-8
EINECS: 241-293-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR