2-(Trifluormethoxy)benzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluormethoxy)benzolsulfonsäureamid

CFF433.2.5 CFF433.10 017450-2.5G 017450-25G 017450-5G
CHMPCFF433.2.5EA 532 EUR
CHMPCFF433.2.5 CHMPCFF433.10 FLUO017450-2.5G FLUO017450-25G FLUO017450-5G
2-(Trifluormethoxy)benzolsulfonamid
2-(Trifluormethoxy)benzolsulfonamid
Formel: C₇H₆F₃NO₃S
Molecular Weight: 241.19 g/mol
MDL: MFCD01320751
CAS-Nummer: 37526-59-3
EINECS: 448-450-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR