2,3-Dichlor-5-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

OS-7457-25G OS-7457-100G
COBBOS-7457-25GEA 37.5 EUR
COBBOS-7457-25G COBBOS-7457-100G
2,3-Dichlor-5-(trifluormethyl)pyridin
2,3-Dichloro-5-(trifluoromethyl)pyridine
Formel: C₆H₂Cl₂F₃N
Molecular Weight: 215.99 g/mol
Siedepunkt: 176
MDL: MFCD00042243
CAS-Nummer: 69045-84-7
EINECS: 410-340-5
UN: 3082
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR