2,3-Dichlorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2647-100G BB-2647-5G BB-2647-25G BB-2647-1G
COBBBB-2647-100GEA 759 EUR
COBBBB-2647-100G COBBBB-2647-5G COBBBB-2647-25G COBBBB-2647-1G
2,3-Dichlorphenylboronsäure
2,3-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190.82 g/mol
MDL: MFCD01075703
CAS-Nummer: 151169-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR