2,3-Difluor-4-hydroxybenzoesäure

Lieferant: COMBI-BLOCKS

CA-4293-5G CA-4293-1G
COBBCA-4293-5GEA 520 EUR
COBBCA-4293-5G COBBCA-4293-1G
2,3-Difluor-4-hydroxybenzoesäure
2,3-Difluoro-4-hydroxybenzoic acid
Formel: C₇H₄F₂O₃
Molecular Weight: 174.1 g/mol
MDL: MFCD01321116
CAS-Nummer: 175968-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR