2,3-Difluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(2,3-difluorophenyl)borane, 2,3-Difluorobenzeneboronic acid, (2,3-Difluorophenyl)metaboric acid

BB-2657-25G BB-2657-100G BB-2657-5G BB-2657-1G
COBBBB-2657-25GEA 0 EUR
COBBBB-2657-25G COBBBB-2657-100G COBBBB-2657-5G COBBBB-2657-1G
2,3-Difluorphenylboronsäure
2,3-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
MDL: MFCD01863170
CAS-Nummer: 121219-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR