2,3-Dimethoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(2,3-dimethoxyphenyl)boran, (2,3-Dimethoxyphenyl)metaborsäure, 2,3-Dimethoxybenzolboronsäure

BB-8359-5G BB-8359-1G
COBBBB-8359-5GEA 50.9 EUR
COBBBB-8359-5G COBBBB-8359-1G
2,3-Dimethoxyphenylboronsäure
2,3-Dimethoxyphenylboronsäure
Formel: C₈H₁₁BO₄
Molecular Weight: 181.98 g/mol
MDL: MFCD02683112
CAS-Nummer: 40972-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR