2,3-Dimethoxypyridin

Lieferant: COMBI-BLOCKS
Achtung

HC-6968-5G HC-6968-25G HC-6968-1G
COBBHC-6968-5GEA 107 EUR
COBBHC-6968-5G COBBHC-6968-25G COBBHC-6968-1G
2,3-Dimethoxypyridin
2,3-Dimethoxypyridine
Formel: C₇H₉NO₂
Molecular Weight: 139,15 g/mol
MDL: MFCD03095043
CAS-Nummer: 52605-97-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR