4-Fluor-2,3-dimethylbenzaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-2,3-dimethylbenzaldehyde

HC-3370-1G HC-3370-5G
COBBHC-3370-1GEA 404 EUR
COBBHC-3370-1G COBBHC-3370-5G
4-Fluor-2,3-dimethylbenzaldehyd
4-Fluor-2,3-dimethylbenzaldehyd
Formel: C₉H₉FO
Molecular Weight: 152,17 g/mol
MDL: MFCD16295095
CAS-Nummer: 363134-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR