2,4,5-Tribrom-1-methylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 2,4,5-Tribrom-1-methyl-1H-imidazol

HI-1030-5G HI-1030-25G HI-1030-100G
COBBHI-1030-5GEA 98.3 EUR
COBBHI-1030-5G COBBHI-1030-25G COBBHI-1030-100G
2,4,5-Tribrom-1-methylimidazol
2,4,5-Tribromo-1-methylimidazole
Formel: C₄H₃Br₃N₂
MDL: MFCD00955564
CAS-Nummer: 1003-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR