2,4,5-Tribromimidazol
Lieferant: FLUOROCHEM
Synonyme:
2,4,5-Tribrom-1H-imidazol
001253-25G
001253-10G
FLUO001253-25GEA
0
EUR
FLUO001253-25G
FLUO001253-10G
2,4,5-Tribromimidazol
2,4,5-Tribromoimidazole
Formel:
C₃HBr₃N₂ Molecular Weight: 304,77 g/mol Siedepunkt: 359 °C (1013 hPa) Schmelzpunkt: 224 °C |
MDL:
MFCD00005184 CAS-Nummer: 2034-22-2 EINECS: 217-997-5 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...